The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_152573.3 | NP_689786.2 | 8 - 9 | 1375 | 144 |
XM_005251730.3 | XP_005251787.1 | 7 - 8 | 1355 | 144 | |
XM_005251731.3 | XP_005251788.1 | 8 - 9 | 1866 | 144 | |
GenBank mRNA | AK290986.1 | - | 8 - 9 | 1117 | 144 |
BC023566.1 | - | 8 - 9 | 1375 | 144 |