Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC35G1
Entrez Gene ID
159371
Gene Name
solute carrier family 35 member G1
Gene Aliases
C10orf60, TMEM20
Chromosome Location
Chr.10: 93893951 - 93914265 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_011539373.2 XP_011537675.1 3 - 4 576 76

View Full Product Details