The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_011545080.2 | XP_011543382.1 | 1 - 2 | 216 | 141 |
XM_017017847.1 | XP_016873336.1 | 1 - 2 | 216 | 141 | |
GenBank mRNA | AK055228.1 | - | 1 - 2 | 176 | 141 |