The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005642.2 | NP_005633.2 | 1 - 1 | 2021 | 123 |
GenBank mRNA | BC032737.1 | - | 1 - 1 | 1979 | 123 |
U18062.1 | - | 1 - 1 | 1957 | 123 | |
X97999.1 | - | 1 - 1 | 2004 | 123 |