The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001008844.2 | NP_001008844.1 | 15 - 16 | 2468 | 118 |
NM_001319034.1 | NP_001305963.1 | 15 - 16 | 2468 | 118 | |
NM_004415.3 | NP_004406.2 | 15 - 16 | 2468 | 118 | |
GenBank mRNA | AB209992.1 | - | 15 - 16 | 2426 | 118 |
AK296756.1 | - | 13 - 14 | 1670 | 118 | |
AK301676.1 | - | 13 - 14 | 1651 | 118 | |
AK304182.1 | - | 15 - 16 | 2371 | 118 | |
BC140802.1 | - | 15 - 16 | 2401 | 118 | |
HM151899.1 | - | 15 - 16 | 2406 | 118 | |
M77830.3 | - | 15 - 16 | 2406 | 118 |