The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005546.3 | NP_005537.3 | 1 - 2 | 218 | 91 |
GenBank mRNA | AB209622.1 | - | 1 - 2 | 271 | 91 |
AK303609.1 | - | 1 - 2 | 184 | 91 | |
AK312846.1 | - | 1 - 2 | 187 | 91 | |
BC109077.2 | - | 1 - 2 | 209 | 91 | |
BC109078.1 | - | 1 - 2 | 209 | 91 |