The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001306075.1 | NP_001293004.1 | 3 - 4 | 775 | 150 |
NM_006527.3 | NP_006518.1 | 4 - 5 | 892 | 150 | |
GenBank mRNA | AK091735.1 | - | 3 - 4 | 352 | 150 |
AK093314.1 | - | 4 - 5 | 365 | 150 | |
AK094257.1 | - | 4 - 5 | 706 | 150 | |
AK315635.1 | - | 4 - 5 | 463 | 150 | |
BC014908.1 | - | 4 - 5 | 461 | 150 | |
BC015703.1 | - | 4 - 5 | 461 | 150 | |
BT007433.1 | - | 4 - 5 | 339 | 150 | |
DB031118.1 | - | 4 - 5 | 365 | 150 | |
U75679.1 | - | 4 - 5 | 454 | 150 | |
Z71188.1 | - | 4 - 5 | 443 | 150 |