The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020728.2 | NP_065779.1 | 8 - 9 | 1134 | 82 |
GenBank mRNA | AB033054.2 | - | 9 - 10 | 920 | 82 |
AK126214.1 | - | 8 - 9 | 779 | 82 | |
AK308824.1 | - | 8 - 9 | 742 | 82 | |
AL833233.1 | - | 8 - 9 | 743 | 82 | |
AY368150.1 | - | 8 - 9 | 1134 | 82 | |
DQ993201.1 | - | 10 - 11 | 1152 | 82 |