The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001290184.1 | NP_001277113.1 | 12 - 13 | 1610 | 97 |
NM_001290186.1 | NP_001277115.1 | 8 - 9 | 1057 | 97 | |
NM_015331.2 | NP_056146.1 | 11 - 12 | 1471 | 97 | |
XM_011509363.2 | XP_011507665.1 | 11 - 12 | 1688 | 97 | |
GenBank mRNA | AF240468.1 | - | 11 - 12 | 1489 | 97 |
AK296153.1 | - | 6 - 7 | 756 | 97 | |
AK314764.1 | - | 11 - 12 | 1378 | 97 | |
AY359120.1 | - | 11 - 12 | 1348 | 97 | |
BC047621.1 | - | 12 - 13 | 1567 | 97 | |
D87442.1 | - | 11 - 12 | 1345 | 97 |