The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001290186.1 | NP_001277115.1 | 1 - 2 | 210 | 109 |
NM_015331.2 | NP_056146.1 | 1 - 2 | 210 | 109 | |
XM_005245053.4 | XP_005245110.1 | 1 - 2 | 427 | 109 | |
XM_011509363.2 | XP_011507665.1 | 1 - 2 | 427 | 109 | |
GenBank mRNA | AF240468.1 | - | 1 - 2 | 228 | 109 |
AK299142.1 | - | 1 - 2 | 97 | 109 | |
AK314764.1 | - | 1 - 2 | 117 | 109 | |
AY359120.1 | - | 1 - 2 | 87 | 109 | |
D87442.1 | - | 1 - 2 | 84 | 109 |