The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003320.4 | NP_003311.2 | 1 - 2 | 396 | 72 |
GenBank mRNA | BC075031.2 | - | 1 - 2 | 188 | 72 |
BC075032.2 | - | 1 - 2 | 188 | 72 | |
U82467.1 | - | 1 - 2 | 396 | 72 |