The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003320.4 | NP_003311.2 | 5 - 6 | 804 | 76 |
NM_177972.2 | NP_813977.1 | 4 - 5 | 547 | 76 | |
XM_005253109.3 | XP_005253166.1 | 4 - 5 | 539 | 76 | |
XM_011520344.2 | XP_011518646.1 | 4 - 5 | 594 | 76 | |
GenBank mRNA | AK293528.1 | - | 4 - 5 | 475 | 76 |
BC075031.2 | - | 5 - 6 | 596 | 76 | |
BC075032.2 | - | 5 - 6 | 596 | 76 | |
U54644.1 | - | 4 - 5 | 550 | 76 | |
U82467.1 | - | 5 - 6 | 804 | 76 |