The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001128148.2 | NP_001121620.1 | 18 - 19 | 2185 | 95 |
NM_001313965.1 | NP_001300894.1 | 17 - 18 | 1983 | 95 | |
NM_001313966.1 | NP_001300895.1 | 16 - 17 | 1787 | 95 | |
NM_003234.3 | NP_003225.2 | 18 - 19 | 2326 | 95 | |
GenBank mRNA | AB209254.1 | - | 18 - 19 | 2089 | 95 |
AK291723.1 | - | 18 - 19 | 2326 | 95 | |
AK294758.1 | - | 16 - 17 | 1717 | 95 | |
BC001188.1 | - | 18 - 19 | 2099 | 95 | |
BX537966.1 | - | 17 - 18 | 1913 | 95 | |
M11507.1 | - | 18 - 19 | 2138 | 95 | |
X01060.1 | - | 18 - 19 | 2306 | 95 |