The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004747.3 | NP_004738.3 | 19 - 20 | 3945 | 71 |
XM_005270276.4 | XP_005270333.1 | 19 - 20 | 3899 | 71 | |
XM_006718056.3 | XP_006718119.1 | 18 - 19 | 2888 | 71 | |
XM_006718057.3 | XP_006718120.1 | 19 - 20 | 3915 | 71 | |
XM_011540341.2 | XP_011538643.1 | 19 - 20 | 4069 | 71 | |
XM_011540342.1 | XP_011538644.1 | 19 - 20 | 3647 | 71 | |
XM_011540344.2 | XP_011538646.1 | 18 - 19 | 3554 | 71 | |
XM_011540345.1 | XP_011538647.1 | 18 - 19 | 3679 | 71 | |
XM_011540346.2 | XP_011538648.1 | 19 - 20 | 3914 | 71 | |
XM_011540347.2 | XP_011538649.1 | 15 - 16 | 3125 | 71 | |
XM_017016913.1 | XP_016872402.1 | 19 - 20 | 3608 | 71 | |
XM_017016914.1 | XP_016872403.1 | 19 - 20 | 3938 | 71 | |
XM_017016915.1 | XP_016872404.1 | 15 - 16 | 3809 | 71 | |
GenBank mRNA | AB011155.2 | - | 19 - 20 | 71 | |
AB091676.1 | - | 17 - 18 | 3410 | 71 | |
AK055172.1 | - | 15 - 16 | 2863 | 71 | |
AK122931.1 | - | 20 - 21 | 3877 | 71 | |
AK314771.1 | - | 3 - 4 | 254 | 71 | |
BC073996.1 | - | 5 - 6 | 655 | 71 | |
BC117695.2 | - | 9 - 10 | 3703 | 71 | |
BC146794.1 | - | 19 - 20 | 71 | ||
BX649131.1 | - | 14 - 15 | 2052 | 71 | |
U61843.1 | - | 4 - 5 | 503 | 71 |