The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001193511.1 | NP_001180440.1 | 3 - 4 | 900 | 73 |
NM_006301.3 | NP_006292.3 | 4 - 5 | 801 | 73 | |
XM_005269138.3 | XP_005269195.1 | 3 - 4 | 828 | 73 | |
XM_005269140.2 | XP_005269197.1 | 3 - 4 | 741 | 73 | |
XM_006719588.3 | XP_006719651.1 | 3 - 4 | 722 | 73 | |
XM_011538725.2 | XP_011537027.1 | 3 - 4 | 1705 | 73 | |
XM_017019956.1 | XP_016875445.1 | 4 - 5 | 729 | 73 | |
GenBank mRNA | AK094195.1 | - | 3 - 4 | 696 | 73 |
AK314530.1 | - | 3 - 4 | 1048 | 73 | |
BC037585.1 | - | 7 - 8 | 73 | ||
BC050050.1 | - | 3 - 4 | 901 | 73 | |
U07358.1 | - | 4 - 5 | 628 | 73 |