Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SLC5A6
Entrez Gene ID
8884
Gene Name
solute carrier family 5 member 6
Gene Aliases
SMVT
Chromosome Location
Chr.2: 27199587 - 27212307 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_021095.2 NP_066918.2 1 - 2 286 61
NR_028323.1 1 - 2 286 61
GenBank mRNA AF069307.1 - 1 - 2 182 61
AF288781.1 - 1 - 2 259 61
AF442149.1 - 1 - 2 222 61
AK027855.1 - 1 - 2 127 61
BI832199.1 - 1 - 2 260 61
DB452476.1 - 1 - 2 286 61

View Full Product Details