The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004949.4 | NP_004940.1 | 10 - 11 | 1973 | 98 |
NM_024422.4 | NP_077740.1 | 10 - 11 | 1973 | 98 | |
XM_005258206.4 | XP_005258263.1 | 10 - 11 | 1474 | 98 | |
GenBank mRNA | AK290313.1 | - | 10 - 11 | 1664 | 98 |
AK297029.1 | - | 11 - 12 | 1633 | 98 | |
BC063291.1 | - | 10 - 11 | 1955 | 98 | |
BX648425.1 | - | 10 - 11 | 1525 | 98 | |
CR749226.1 | - | 10 - 11 | 1692 | 98 | |
EF017811.1 | - | 10 - 11 | 1512 | 98 | |
EF017812.1 | - | 10 - 11 | 1512 | 98 | |
X56807.1 | - | 10 - 11 | 1552 | 98 |