The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004949.4 | NP_004940.1 | 13 - 14 | 2586 | 61 |
NM_024422.4 | NP_077740.1 | 13 - 14 | 2586 | 61 | |
XM_005258206.4 | XP_005258263.1 | 13 - 14 | 2087 | 61 | |
GenBank mRNA | AK290313.1 | - | 13 - 14 | 2277 | 61 |
AK297029.1 | - | 14 - 15 | 2246 | 61 | |
BC063291.1 | - | 13 - 14 | 2568 | 61 | |
BX648425.1 | - | 13 - 14 | 2138 | 61 | |
CR749226.1 | - | 13 - 14 | 2305 | 61 | |
EF017811.1 | - | 13 - 14 | 2125 | 61 | |
EF017812.1 | - | 13 - 14 | 2125 | 61 | |
X56807.1 | - | 13 - 14 | 2165 | 61 |