Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FUCA1
Entrez Gene ID
2517
Gene Name
fucosidase, alpha-L- 1, tissue
Gene Aliases
FUCA
Chromosome Location
Chr.1: 23845077 - 23868369 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000147.4 NP_000138.2 5 - 6 1051 82
XM_005245821.2 XP_005245878.1 5 - 6 1188 82
XM_011541167.2 XP_011539469.1 5 - 6 636 82
XM_017000905.1 XP_016856394.1 5 - 6 733 82
GenBank mRNA AB074175.1 - 5 - 6 972 82
AK092914.1 - 6 - 7 906 82
AK095443.1 - 6 - 7 923 82
AK095455.1 - 6 - 7 937 82
AK225468.1 - 5 - 6 972 82
AK301302.1 - 7 - 8 901 82
AK314649.1 - 5 - 6 972 82
BC017338.2 - 5 - 6 974 82
M10355.1 - 5 - 6 746 82
M29877.1 - 5 - 6 971 82
X01390.1 - 5 - 6 746 82

View Full Product Details