Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LIN7A
Entrez Gene ID
8825
Gene Name
lin-7 homolog A, crumbs cell polarity complex component
Gene Aliases
LIN-7A, LIN7, MALS-1, TIP-33, VELI1
Chromosome Location
Chr.12: 80792520 - 80937915 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004664.3 NP_004655.1 2 - 3 386 107
NR_136888.1 2 - 3 386 107
XM_011538928.2 XP_011537230.1 2 - 3 380 107
GenBank mRNA AF028826.1 - 1 - 2 102 107
AF087693.1 - 2 - 3 355 107
AF173081.1 - 2 - 3 386 107
AK309256.1 - 2 - 3 175 107
AK315321.1 - 2 - 3 230 107
BC062332.1 - 2 - 3 251 107
BC099921.1 - 2 - 3 196 107
BC118609.1 - 2 - 3 330 107
BC122561.1 - 2 - 3 330 107
CR407680.1 - 2 - 3 193 107

View Full Product Details