Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.



Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA AK000013.1 - 1 1146 Chr. 16 - 66549280 - 66551212 [+] on Build GRCh38 72


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream brain expressed, associated with NEDD4, 1 BEAN1 BEAN, SCA31 -55.8kb Sense Chr. - 66427297 - 66495288 on Build GRCh38
Downstream chemokine-like factor CKLF C32, CKLF1, CKLF2, CKLF3, CKLF4, HSPC224, UCK-1 1.4kb Sense Chr. - 66552563 - 66566287 on Build GRCh38

View Full Product Details