The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001321299.1 | NP_001308228.1 | 16 - 17 | 3610 | 67 |
NM_032387.4 | NP_115763.2 | 17 - 18 | 3699 | 67 | |
XM_011525135.2 | XP_011523437.1 | 17 - 18 | 4629 | 67 | |
XM_017024962.1 | XP_016880451.1 | 17 - 18 | 4629 | 67 | |
XM_017024963.1 | XP_016880452.1 | 17 - 18 | 4626 | 67 | |
XM_017024964.1 | XP_016880453.1 | 16 - 17 | 4491 | 67 | |
XM_017024965.1 | XP_016880454.1 | 16 - 17 | 4488 | 67 | |
XM_017024966.1 | XP_016880455.1 | 14 - 15 | 2751 | 67 | |
GenBank mRNA | AF390018.1 | - | 17 - 18 | 3652 | 67 |
AJ316534.1 | - | 17 - 18 | 3648 | 67 | |
BC136664.1 | - | 17 - 18 | 3649 | 67 | |
BM683764.1 | - | 2 - 3 | 67 |