The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001099339.1 | NP_001092809.1 | 7 - 8 | 1088 | 110 |
NM_053041.2 | NP_444269.2 | 7 - 8 | 1091 | 110 | |
GenBank mRNA | AK027893.1 | - | 7 - 8 | 538 | 110 |
AK096307.1 | - | 7 - 8 | 1080 | 110 | |
AY542162.1 | - | 7 - 8 | 474 | 110 | |
BC000628.2 | - | 7 - 8 | 524 | 110 | |
BC047440.1 | - | 7 - 8 | 652 | 110 |