Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ATG2B
Entrez Gene ID
55102
Gene Name
autophagy related 2B
Gene Aliases
C14orf103
Chromosome Location
Chr.14: 96281258 - 96363341 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006720187.2 XP_006720250.1 37 - 38 5859 95
XM_017021397.1 XP_016876886.1 37 - 38 5862 95
GenBank mRNA AK001104.1 - 9 - 10 1260 95
AK128275.1 - 18 - 19 2307 95
AK302641.1 - 10 - 11 1483 95
BC015016.2 - 11 - 12 1590 95

View Full Product Details