The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_006720187.2 | XP_006720250.1 | 6 - 7 | 1292 | 103 |
XM_017021397.1 | XP_016876886.1 | 6 - 7 | 1292 | 103 | |
GenBank mRNA | AK055200.1 | - | 6 - 7 | 1292 | 103 |