The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001280542.1 | NP_001267471.1 | 1 - 2 | 59 | 76 |
NM_012074.4 | NP_036206.3 | 1 - 2 | 59 | 76 | |
XM_017021670.1 | XP_016877159.1 | 1 - 2 | 59 | 76 | |
GenBank mRNA | AK024141.1 | - | 1 - 2 | 44 | 76 |
BC026305.1 | - | 1 - 2 | 44 | 76 | |
BC060801.1 | - | 1 - 2 | 44 | 76 | |
DA010060.1 | - | 1 - 2 | 45 | 76 | |
DA407279.1 | - | 1 - 2 | 80 | 76 | |
DB140241.1 | - | 1 - 2 | 59 | 76 |