Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RGN
Entrez Gene ID
9104
Gene Name
regucalcin
Gene Aliases
GNL, HEL-S-41, RC, SMP30
Chromosome Location
Chr.X: 47078299 - 47093314 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001282848.1 NP_001269777.1 6 - 7 1636 105
NM_001282849.1 NP_001269778.1 5 - 6 1469 105
NM_004683.5 NP_004674.1 5 - 6 1065 105
NM_152869.3 NP_690608.1 6 - 7 1685 105
XM_006724567.2 XP_006724630.1 4 - 5 785 105
GenBank mRNA AB028125.1 - 5 - 6 789 105
AB032064.1 - 5 - 6 847 105
AK223360.1 - 5 - 6 1065 105
AK290136.1 - 5 - 6 1003 105
BC050371.1 - 5 - 6 1407 105
BC073173.1 - 5 - 6 879 105
D31815.1 - 5 - 6 788 105
EU794655.1 - 5 - 6 847 105

View Full Product Details