The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001324029.1 | NP_001310958.1 | 11 - 12 | 1512 | 67 |
NM_001324030.1 | NP_001310959.1 | 12 - 13 | 1694 | 67 | |
NM_001324031.1 | NP_001310960.1 | 13 - 14 | 1880 | 67 | |
NM_001324033.1 | NP_001310962.1 | 13 - 14 | 1966 | 67 | |
NM_001324034.1 | NP_001310963.1 | 12 - 13 | 1968 | 67 | |
NM_020210.3 | NP_064595.2 | 13 - 14 | 1972 | 67 | |
NM_198925.2 | NP_945119.1 | 12 - 13 | 1938 | 67 | |
GenBank mRNA | AB051532.1 | - | 12 - 13 | 1926 | 67 |
AK026108.1 | - | 12 - 13 | 1671 | 67 | |
AK026133.1 | - | 5 - 6 | 842 | 67 | |
AK289991.1 | - | 13 - 14 | 1941 | 67 | |
AL390081.1 | - | 8 - 9 | 1198 | 67 | |
AY358392.1 | - | 13 - 14 | 1929 | 67 | |
AY445887.1 | - | 12 - 13 | 1949 | 67 | |
BC017658.1 | - | 4 - 5 | 552 | 67 | |
BC094849.1 | - | 5 - 6 | 742 | 67 | |
BC110853.1 | - | 12 - 13 | 1693 | 67 | |
BC126438.1 | - | 12 - 13 | 1847 | 67 | |
BC144141.1 | - | 13 - 14 | 1875 | 67 | |
BX537671.1 | - | 9 - 10 | 1891 | 67 |