The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001145400.1 | NP_001138872.1 | 9 - 10 | 1767 | 81 |
NM_139174.3 | NP_631913.3 | 10 - 11 | 2013 | 81 | |
XM_005255814.1 | XP_005255871.1 | 9 - 10 | 1797 | 81 | |
XM_011522919.1 | XP_011521221.1 | 10 - 11 | 1983 | 81 | |
XM_011522920.1 | XP_011521222.1 | 10 - 11 | 1708 | 81 | |
GenBank mRNA | AF447586.1 | - | 9 - 10 | 1705 | 81 |
AK093046.1 | - | 10 - 11 | 1992 | 81 | |
AK093523.1 | - | 7 - 8 | 2267 | 81 | |
AK315171.1 | - | 9 - 10 | 1686 | 81 | |
BC033491.1 | - | 9 - 10 | 1705 | 81 | |
BU731201.1 | - | 1 - 2 | 81 |