Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LHFPL2
Entrez Gene ID
10184
Gene Name
lipoma HMGIC fusion partner-like 2
Gene Aliases
-
Chromosome Location
Chr.5: 78485215 - 78648895 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005779.2 NP_005770.1 3 - 4 498 92
XM_006714515.1 XP_006714578.1 2 - 3 185 92
GenBank mRNA CN314535.1 - 3 - 4 322 92
DA094988.1 - 3 - 4 448 92

View Full Product Details