Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NTM
Entrez Gene ID
50863
Gene Name
neurotrimin
Gene Aliases
HNT, IGLON2, NTRI
Chromosome Location
Chr.11: 131370326 - 132336822 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001048209.1 NP_001041674.1 3 - 4 732 119
NM_001144058.1 NP_001137530.1 2 - 3 1065 119
NM_001144059.1 NP_001137531.1 2 - 3 1065 119
NM_016522.2 NP_057606.1 2 - 3 1065 119
XM_005271580.3 XP_005271637.1 3 - 4 882 119
XM_005271581.3 XP_005271638.1 2 - 3 835 119
XM_005271582.3 XP_005271639.1 3 - 4 882 119
XM_006718855.3 XP_006718918.1 4 - 5 989 119
XM_006718857.3 XP_006718920.1 3 - 4 909 119
XM_011542861.2 XP_011541163.1 2 - 3 835 119
XM_017017851.1 XP_016873340.1 2 - 3 835 119
XM_017017852.1 XP_016873341.1 3 - 4 882 119
XM_017017853.1 XP_016873342.1 4 - 5 989 119
XM_017017854.1 XP_016873343.1 2 - 3 835 119
XM_017017855.1 XP_016873344.1 3 - 4 882 119
XM_017017856.1 XP_016873345.1 4 - 5 595 119
XM_017017857.1 XP_016873346.1 4 - 5 886 119
XM_017017858.1 XP_016873347.1 4 - 5 879 119
XM_017017859.1 XP_016873348.1 4 - 5 595 119
XM_017017860.1 XP_016873349.1 3 - 4 912 119
XM_017017861.1 XP_016873350.1 4 - 5 989 119
XM_017017862.1 XP_016873351.1 4 - 5 876 119
XM_017017863.1 XP_016873352.1 3 - 4 912 119
XM_017017864.1 XP_016873353.1 3 - 4 880 119
XM_017017865.1 XP_016873354.1 3 - 4 882 119
XM_017017866.1 XP_016873355.1 3 - 4 909 119
GenBank mRNA AF126426.1 - 2 - 3 665 119
AK293448.1 - 3 - 4 700 119
AK293487.1 - 4 - 5 681 119
AK294137.1 - 3 - 4 611 119
AK315964.1 - 3 - 4 727 119
AY358331.1 - 3 - 4 534 119
BC050716.1 - 2 - 3 415 119
BE263639.1 - 1 - 2 131 119
EF043567.1 - 2 - 3 421 119

View Full Product Details