The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001304536.1 | NP_001291465.1 | 11 - 12 | 2162 | 70 |
NM_001304537.1 | NP_001291466.1 | 9 - 10 | 1929 | 70 | |
NM_004438.4 | NP_004429.1 | 10 - 11 | 1997 | 70 | |
XM_005246374.2 | XP_005246431.1 | 10 - 11 | 1997 | 70 | |
GenBank mRNA | AK290306.1 | - | 11 - 12 | 70 | |
AK300772.1 | - | 9 - 10 | 1867 | 70 | |
AK312380.1 | - | 10 - 11 | 1943 | 70 | |
BC026327.1 | - | 10 - 11 | 1922 | 70 | |
BC105000.1 | - | 10 - 11 | 1923 | 70 | |
BC105002.1 | - | 10 - 11 | 1923 | 70 | |
L36645.1 | - | 10 - 11 | 1920 | 70 |