The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001304536.1 | NP_001291465.1 | 16 - 17 | 2960 | 100 |
NM_001304537.1 | NP_001291466.1 | 14 - 15 | 2727 | 100 | |
NM_004438.4 | NP_004429.1 | 15 - 16 | 2795 | 100 | |
XM_005246374.2 | XP_005246431.1 | 15 - 16 | 2795 | 100 | |
GenBank mRNA | AK290306.1 | - | 16 - 17 | 2960 | 100 |
AK300772.1 | - | 14 - 15 | 2665 | 100 | |
AK312380.1 | - | 15 - 16 | 2741 | 100 | |
BC026327.1 | - | 15 - 16 | 2720 | 100 | |
BC105000.1 | - | 15 - 16 | 2721 | 100 | |
BC105002.1 | - | 15 - 16 | 2721 | 100 | |
BQ009361.1 | - | 2 - 3 | 100 | ||
L36645.1 | - | 15 - 16 | 2718 | 100 |