The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001304536.1 | NP_001291465.1 | 9 - 10 | 1989 | 73 |
NM_001304537.1 | NP_001291466.1 | 7 - 8 | 1756 | 73 | |
NM_004438.4 | NP_004429.1 | 8 - 9 | 1824 | 73 | |
XM_005246374.2 | XP_005246431.1 | 8 - 9 | 1824 | 73 | |
GenBank mRNA | AK290306.1 | - | 9 - 10 | 1989 | 73 |
AK300772.1 | - | 7 - 8 | 1694 | 73 | |
AK312380.1 | - | 8 - 9 | 1770 | 73 | |
BC026327.1 | - | 8 - 9 | 1749 | 73 | |
BC105000.1 | - | 8 - 9 | 1750 | 73 | |
BC105002.1 | - | 8 - 9 | 1750 | 73 | |
L36645.1 | - | 8 - 9 | 1747 | 73 |