The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014799.3 | NP_055614.1 | 1 - 2 | 245 | 80 |
GenBank mRNA | AB014598.2 | - | 1 - 2 | 208 | 80 |