Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CXorf56
Entrez Gene ID
63932
Gene Name
chromosome X open reading frame 56
Gene Aliases
-
Chromosome Location
Chr.X: 119538149 - 119565434 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001170569.1 NP_001164040.1 1 - 2 289 61
GenBank mRNA AK096297.1 - 1 - 2 253 61
DA590933.1 - 1 - 2 291 61

View Full Product Details