The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001178032.2 | NP_001171503.1 | 2 - 3 | 545 | 73 |
NM_001178033.2 | NP_001171504.1 | 2 - 3 | 383 | 73 | |
NM_001243804.1 | NP_001230733.1 | 2 - 3 | 290 | 73 | |
NM_018684.3 | NP_061154.1 | 2 - 3 | 383 | 73 | |
NR_045044.1 | 2 - 3 | 638 | 73 | ||
GenBank mRNA | AB032992.1 | - | 2 - 3 | 383 | 73 |
AF270491.1 | - | 2 - 3 | 315 | 73 | |
AK022807.1 | - | 2 - 3 | 514 | 73 | |
AK123763.1 | - | 3 - 4 | 776 | 73 | |
AK293570.1 | - | 2 - 3 | 300 | 73 | |
AK315485.1 | - | 2 - 3 | 298 | 73 | |
BC004411.1 | - | 2 - 3 | 334 | 73 | |
BI770655.1 | - | 2 - 3 | 467 | 73 | |
BP376634.1 | - | 2 - 3 | 290 | 73 | |
CX782941.1 | - | 2 - 3 | 255 | 73 |