Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZC4H2
Entrez Gene ID
55906
Gene Name
zinc finger C4H2-type containing
Gene Aliases
HCA127, KIAA1166, WRWF, WWS
Chromosome Location
Chr.X: 64915802 - 65034744 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001178032.2 NP_001171503.1 4 - 5 881 72
NM_001243804.1 NP_001230733.1 4 - 5 626 72
NM_018684.3 NP_061154.1 4 - 5 719 72
NR_045044.1 4 - 5 974 72
GenBank mRNA AF270491.1 - 4 - 5 651 72
AK001547.1 - 1 - 2 148 72
AK022807.1 - 4 - 5 850 72
AK022918.1 - 4 - 5 613 72
AK123763.1 - 5 - 6 1112 72
AK222708.1 - 4 - 5 636 72
AK315485.1 - 4 - 5 634 72
BC004411.1 - 4 - 5 670 72
CX782941.1 - 4 - 5 591 72

View Full Product Details