Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NFYA
Entrez Gene ID
4800
Gene Name
nuclear transcription factor Y subunit alpha
Gene Aliases
CBF-A, CBF-B, HAP2, NF-YA
Chromosome Location
Chr.6: 41072968 - 41102407 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002505.4 NP_002496.1 5 - 6 622 82
NM_021705.3 NP_068351.1 4 - 5 535 82
GenBank mRNA BC039244.1 - 4 - 5 788 82
BM561405.1 - 4 - 5 535 82
GQ167198.1 - 4 - 5 441 82
KU159426.1 - 4 - 5 597 82
M59079.1 - 2 - 3 207 82
X59711.1 - 5 - 6 618 82

View Full Product Details