Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TFRC
Entrez Gene ID
7037
Gene Name
transferrin receptor
Gene Aliases
CD71, IMD46, T9, TFR, TFR1, TR, TRFR, p90
Chromosome Location
Chr.3: 196049284 - 196082161 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001128148.2 NP_001121620.1 1 - 2 121 81
NM_001313965.1 NP_001300894.1 1 - 2 121 81
NM_001313966.1 NP_001300895.1 1 - 2 121 81
GenBank mRNA CF994006.1 - 1 - 2 149 81
DA460386.1 - 1 - 2 121 81
M11507.1 - 1 - 2 74 81

View Full Product Details