The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014573.2 | NP_055388.2 | 2 - 3 | 423 | 66 |
XM_005257965.3 | XP_005258022.1 | 2 - 3 | 472 | 66 | |
GenBank mRNA | AK293331.1 | - | 2 - 3 | 278 | 66 |
AK299511.1 | - | 2 - 3 | 316 | 66 | |
AK307678.1 | - | 2 - 3 | 315 | 66 | |
BC017362.2 | - | 2 - 3 | 294 | 66 | |
BC045655.1 | - | 3 - 4 | 432 | 66 | |
BC091504.1 | - | 2 - 3 | 294 | 66 | |
DA410183.1 | - | 2 - 3 | 423 | 66 | |
L19183.1 | - | 2 - 3 | 317 | 66 |