The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001291866.1 | NP_001278795.1 | 7 - 8 | 890 | 140 |
NM_004481.4 | NP_004472.1 | 7 - 8 | 812 | 140 | |
XM_017000963.1 | XP_016856452.1 | 7 - 8 | 772 | 140 | |
XM_017000964.1 | XP_016856453.1 | 7 - 8 | 1397 | 140 | |
XM_017000965.1 | XP_016856454.1 | 7 - 8 | 890 | 140 | |
XM_017000966.1 | XP_016856455.1 | 7 - 8 | 715 | 140 | |
GenBank mRNA | AK290048.1 | - | 7 - 8 | 738 | 140 |
AK296886.1 | - | 4 - 5 | 412 | 140 | |
AK300453.1 | - | 7 - 8 | 890 | 140 | |
AK304029.1 | - | 6 - 7 | 642 | 140 | |
BC041120.1 | - | 7 - 8 | 798 | 140 | |
X85019.1 | - | 7 - 8 | 726 | 140 |