The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017677.3 | NP_060147.2 | 1 - 2 | 120 | 91 |
GenBank mRNA | BC012399.1 | - | 1 - 2 | 98 | 91 |
DA691850.1 | - | 1 - 2 | 120 | 91 |