The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001194937.1 | NP_001181866.1 | 16 - 17 | 2715 | 68 |
NM_001194938.1 | NP_001181867.1 | 15 - 16 | 2556 | 68 | |
NM_015100.3 | NP_055915.2 | 16 - 17 | 2742 | 68 | |
NM_145796.3 | NP_665739.3 | 14 - 15 | 2457 | 68 | |
NM_207171.2 | NP_997054.1 | 14 - 15 | 2269 | 68 | |
XM_005244999.2 | XP_005245056.1 | 16 - 17 | 5410 | 68 | |
XM_005245000.4 | XP_005245057.1 | 16 - 17 | 3301 | 68 | |
XM_005245001.2 | XP_005245058.1 | 16 - 17 | 2526 | 68 | |
XM_005245005.1 | XP_005245062.1 | 15 - 16 | 2305 | 68 | |
XM_005245006.4 | XP_005245063.1 | 15 - 16 | 3221 | 68 | |
XM_011509331.2 | XP_011507633.1 | 14 - 15 | 2318 | 68 | |
XM_017000744.1 | XP_016856233.1 | 16 - 17 | 2544 | 68 | |
XM_017000745.1 | XP_016856234.1 | 16 - 17 | 5384 | 68 | |
XM_017000746.1 | XP_016856235.1 | 16 - 17 | 3275 | 68 | |
XM_017000747.1 | XP_016856236.1 | 14 - 15 | 2281 | 68 | |
XM_017000748.1 | XP_016856237.1 | 15 - 16 | 2288 | 68 | |
XM_017000749.1 | XP_016856238.1 | 15 - 16 | 2659 | 68 | |
GenBank mRNA | AB007930.1 | - | 14 - 15 | 2264 | 68 |
AB037911.1 | - | 15 - 16 | 2324 | 68 | |
AB075477.1 | - | 15 - 16 | 2324 | 68 | |
AK299791.1 | - | 10 - 11 | 1241 | 68 | |
AK300307.1 | - | 16 - 17 | 2429 | 68 | |
AK302501.1 | - | 15 - 16 | 2306 | 68 | |
BX537561.1 | - | 14 - 15 | 2171 | 68 | |
BX537838.1 | - | 16 - 17 | 2743 | 68 | |
BX649163.1 | - | 15 - 16 | 2297 | 68 |