The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016458.3 | NP_057542.2 | 1 - 2 | 660 | 90 |
GenBank mRNA | AK098226.1 | - | 2 - 3 | 354 | 90 |
BC003035.1 | - | 1 - 2 | 660 | 90 | |
BC009915.2 | - | 1 - 2 | 660 | 90 | |
BC015471.1 | - | 2 - 3 | 191 | 90 |