The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018132.3 | NP_060602.2 | 4 - 5 | 362 | 139 |
XM_005249205.1 | XP_005249262.1 | 4 - 5 | 423 | 139 | |
XM_011514723.2 | XP_011513025.2 | 3 - 4 | 298 | 139 | |
GenBank mRNA | AK293016.1 | - | 4 - 5 | 367 | 139 |
BC016279.1 | - | 4 - 5 | 352 | 139 | |
BI768787.1 | - | 4 - 5 | 380 | 140 |