The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018132.3 | NP_060602.2 | 1 - 2 | 67 | 69 |
GenBank mRNA | AK293016.1 | - | 1 - 2 | 72 | 69 |
BC016279.1 | - | 1 - 2 | 57 | 69 | |
BI768787.1 | - | 1 - 2 | 85 | 69 |