Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GPBP1
Entrez Gene ID
65056
Gene Name
GC-rich promoter binding protein 1
Gene Aliases
GPBP, SSH6, VASCULIN
Chromosome Location
Chr.5: 57173826 - 57264679 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001127235.2 NP_001120707.2 1 - 2 955 76
NM_022913.3 NP_075064.1 2 - 3 1219 76
XM_017009754.1 XP_016865243.1 1 - 2 627 76
XM_017009755.1 XP_016865244.1 1 - 2 627 76
XM_017009756.1 XP_016865245.1 1 - 2 627 76
GenBank mRNA AY226828.1 - 2 - 3 1050 76
BC000267.1 - 1 - 2 457 76
BC113004.2 - 1 - 2 968 76

View Full Product Details