The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001042529.2 | NP_001035994.1 | 9 - 10 | 1792 | 67 |
NM_001042532.3 | NP_001035997.2 | 10 - 11 | 1924 | 67 | |
NM_025233.6 | NP_079509.5 | 8 - 9 | 2184 | 67 | |
XM_006722116.3 | XP_006722179.1 | 9 - 10 | 1818 | 67 | |
XM_011525300.1 | XP_011523602.1 | 9 - 10 | 1988 | 67 | |
XM_017025167.1 | XP_016880656.1 | 6 - 7 | 1873 | 67 | |
XM_017025168.1 | XP_016880657.1 | 9 - 10 | 1397 | 67 | |
GenBank mRNA | AF208536.1 | - | 8 - 9 | 1144 | 67 |
AF453478.1 | - | 8 - 9 | 1635 | 67 | |
AK075415.1 | - | 8 - 9 | 2092 | 67 | |
AK092498.1 | - | 6 - 7 | 1859 | 67 | |
AK225485.1 | - | 9 - 10 | 1721 | 67 | |
AK297153.1 | - | 10 - 11 | 1847 | 67 | |
AK314076.1 | - | 10 - 11 | 1848 | 67 | |
AL360167.1 | - | 8 - 9 | 1144 | 67 | |
AL833588.1 | - | 9 - 10 | 1716 | 67 | |
AY094602.1 | - | 9 - 10 | 1745 | 67 | |
BC006354.1 | - | 7 - 8 | 885 | 67 | |
BC020985.1 | - | 8 - 9 | 1360 | 67 | |
BC067254.1 | - | 8 - 9 | 2088 | 67 | |
BT007168.1 | - | 6 - 7 | 618 | 67 | |
U18919.1 | - | 6 - 7 | 643 | 67 |