The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005557.3 | NP_005548.2 | 1 - 2 | 668 | 73 |
GenBank mRNA | AF061812.1 | - | 1 - 2 | 543 | 73 |
AK290853.1 | - | 1 - 2 | 606 | 73 | |
BC039169.1 | - | 1 - 2 | 579 | 73 | |
S79867.1 | - | 1 - 2 | 529 | 73 |